missing translation for 'onlineSavingsMsg'
Learn More

Anemarrhenasaponin I, MedChemExpress

Product Code. 16481738
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481738 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481738 Supplier Medchem Express Supplier No. HYN4213
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Anemarrhenasaponin I, a traditional Chinese medicine, shows remarkable inhibiting effect on platelet aggregation.

Anemarrhenasaponin I, a traditional Chinese medicine, shows remarkable inhibiting effect on platelet aggregation.

Specifications

Chemical Name or Material Anemarrhenasaponin I
CAS 163047-21-0
Color White
Purity Grade Notes Research
Molecular Formula C39H66O14
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (65.88 mM)
SMILES C[C@@]12[C@]([C@@H](O)[C@@]3([H])[C@]2([H])[C@@H](C(CCC(C)C)(O)O3)C)([H])[C@@]4([H])[C@]([C@@]5([C@](C[C@@H](O[C@@]6([H])[C@@H]([C@H]([C@@H](O)[C@@H](CO)O6)O)O[C@]7([H])O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)CO)CC5)([H])CC4)C)([H])CC1
Molecular Weight (g/mol) 758.93
Formula Weight 758.93
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.