missing translation for 'onlineSavingsMsg'
Learn More

Androsterone, Mikromol™

Product Code. 17953966
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953966 100 mg 100mg
17901256 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953966 Supplier Mikromol Supplier No. MM0323.16
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Androsterone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Androsterone
CAS 53-41-8
Analyte or Component Names Androsterone
Shipping Condition Room Temperature
Molecular Formula C19 H30 O2
Certifications/Compliance ISO 17025
Synonym Androsterone (8CI), 3-Epihydroxyetioallocholan-17-one, Androkinin, Androtin, Atromide ICI, NSC 9898, U 60366, 3alpha-Hydroxy-17-androstanone, 3alpha-Hydroxy-5alpha-androstan-17-one, 3alpha-Hydroxy-5alpha-androstane-17-one, 3alpha-Hydroxyetioallocholan-17-one, 5alpha-Androstan-17-one-3alpha-ol, 5alpha-Androstan-3alpha-ol-17-one, 5alpha-Androstane-3alpha-ol-17-one, 5alpha-Androsterone, Androstan-17-one, 3-hydroxy-, (3alpha,5alpha)-
InChI Formula InChI=1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18-,19-/m0/s1
SMILES C[C@]12CC[C@@H](O)C[C@@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O
IUPAC Name (3R,5S,8R,9S,10S,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 290.44
Quantity 100 mg
Formula Weight 290.2246 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.