missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Androstenedione Competitive ELISA Kit
Description
Androstenedione is a steroid hormone also known as androst-4-ene-3,17-dione. It is synthesized from dehydroepiandrosterone (DHEA) or 17-hydroxyprogesterone. By itself, androstenedione is a weak androgen though it is a precursor for biosynthesis of stronger androgens, such as testosterone and estrogens. Therefore, androstenedione is classified as a controlled drug and a supplement to enhance athletic performance and body energy.
Specifications
Specifications
| Assay Range | 5 to 1200 pg/mL |
| Assay Sensitivity | 2.3 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Saliva, Serum, Urine |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Interassay CV | <10% |
| Intraassay CV | <10 |
| Kit Contents | Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers |
| Label or Dye | HRP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?