missing translation for 'onlineSavingsMsg'
Learn More

Andropanolide, MedChemExpress

Product Code. 16431658
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431658 5 mg 5mg
16422579 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431658 Supplier Medchem Express Supplier No. HYN1912
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Andrographolide (Andro) is a small antagonist for NF-κB activation by covalent modifying reduced cysteine 62 of p50.

Andrographolide is a bicyclic diterpenoid lactone mainly produced from the plant Andrographis (Andrographis paniculate). Andrographolide suppresses the activation of NF-κB in stimulated endothelial cells, which reduces the expression of cell adhesion molecule E-selectin and prevents E-selectin-mediated leukocyte adhesion, but has no effect on IκBα degradation, p50 and p65 nuclear translocation.

Specifications

Chemical Name or Material Andropanolide
CAS 869807-57-8
Color White
Purity Grade Notes Research
Molecular Formula C20H30O5
Quantity 5 mg
SMILES O=C1OC[C@@H](O)/C1=C/C[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 98.78%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.