missing translation for 'onlineSavingsMsg'
Learn More

Andrographolide, MedChemExpress

Product Code. 16442366
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442366

Brand: Medchem Express HYN0191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Andrographolide is a NF-κB inhibitor, which inhibits NF-κB activation through covalent modification of a cysteine residue on p50 in endothelial cells without affecting IκBα degradation or p50/p65 nuclear translocation. Andrographolide has antiviral effects.

Andrographolide is a NF-κB inhibitor, which inhibits NF-κB activation through covalent modification of a cysteine residue on p50 in endothelial cells without affecting IκBα degradation or p50/p65 nuclear translocation. Andrographolide has antiviral effects.

Specifications

Chemical Name or Material Andrographolide
Physical Form Solid
CAS 5508-58-7
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C20H30O5
For Use With (Application) Cancer-programmed cell death
Synonym Andrographis
Solubility Information DMSO : 100 mg/mL (285.35 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1OC[C@@H](O)/C1=C\C[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 97.46%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.