missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Androgen Receptor, Ligand Binding Domain, GST Tag, His Tag Recombinant Protein
Rat Androgen Receptor, Ligand Binding Domain, GST Tag, His Tag Recombinant Protein
Brand: Invitrogen A15675
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Androgen receptor ligand binding domain (AR-LBD) is a 64.42 kDa recombinant rat protein, expressed as a GST- and His-tagged protein in insect cells. The protein contains the hinge region and an LBD amino acid sequence identical to that of the human LBD. Differences in the rat and human hinge region are present at human amino acids 646, 649, 650, 655, 656, 657, and 660. AR-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand-binding and coregulator recruitment studies.AR-LBD rat recombinant protein is:
Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
Specifications
NP_036634 | |
2 mg | |
AR | |
Recombinant | |
Store in ultra-cold freezer (-68 to -85°C). | |
Growth Factors & Receptors |
24208.0 | |
Tfm, Andr | |
Rat | |
GST Tag, His Tag | |
Baculovirus | |
Ligand Binding Domain, Recombinant |
For Research Use Only. Not for use in diagnostic procedures.