missing translation for 'onlineSavingsMsg'
Learn More

Androgen R/NR3C4, p Ser210, p Ser213 Antibody (156C135.2) - Azide and BSA Free, Novus Biologicals™

Product Code. 18419971 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419971 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419971 Supplier Novus Biologicals Supplier No. NBP2310991.0mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Androgen R/NR3C4 (p Ser210, p Ser213)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 156C135.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-4 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000
Formulation PBS with No Preservative
Gene Accession No. P10275
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols AR
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Purification Method Protein G purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 367
Test Specificity In IGF-1 stimulated LNCaP cells (passage number 38), a ∼110 kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen.
Target Species Human, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.