Learn More
Androgen R/NR3C4 Antibody (CL9255) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88922APC
Product Code. 30307036
  • £574.00 / 0.10mL
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Androgen R/NR3C4
Monoclonal
APC
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
This antibody was developed using a synthetic peptide derived from P10275, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Cellular Markers, Phospho Specific
367
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9255
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified