missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4 Antibody (CL9255) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88922AF350
This item is not returnable.
View return policy
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Androgen R/NR3C4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| This antibody was developed using a synthetic peptide derived from P10275, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 367 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9255 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction