missing translation for 'onlineSavingsMsg'
Learn More

Ancient ubiquitous protein 1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18424434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424434 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424434 Supplier Novus Biologicals Supplier No. NBP149916AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ancient ubiquitous protein 1 Polyclonal antibody specifically detects Ancient ubiquitous protein 1 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ancient ubiquitous protein 1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias ancient ubiquitous protein 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 426 and 476 of human Ancient Ubiquitous Protein 1 using the numbering given in entry NP_853553.1 (GeneID 550).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 550
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.