missing translation for 'onlineSavingsMsg'
Learn More

(±)-Anatoxin A fumarate, Tocris Bioscience™

Product Code. 30001130
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001130 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001130 Supplier Tocris Bioscience Supplier No. 0789/1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nicotinic agonist

(±)-Anatoxin A fumarate is a potent nicotinic agonist (Ki values are 1.25 and 1840 nM for α4β2 and α7 nicotinic receptors respectively). (?)-Anatoxin A fumarate stimulates [3H]-dopamine release from rat striatal synaptosomes (EC50 = 136 nM). (?)-Anatoxin A fumarate induces apoptosis in rat thymocytes and Vero cells by generation of reactive oxygen species and caspase activation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 122564-82-3
Molecular Formula C10H15NO·C4H4O4
Quantity 1 mg
Synonym (±)-2-Acetyl-9-aza bicyclo[4.2.1]non-2-ene fumarate
Formula Weight 281.31
Percent Purity >96%
Chemical Name or Material (±)-Anatoxin A fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.