Learn More
ABEBA™ Anatom Green/Yellow Safety Shoes S3 ESD
Product Code. 17101235
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 17101235
Brand: ABEBA™ 32251/52
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Green/yellow Anatom shoes made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
  • Comfortable, robust, durable shoes
  • Comply to EN ISO 20345: 2012, S3, SRC
  • ESD dissipative according to EN 61340 class climatic 1
  • Slip resistant PU double density outsole
  • Green/yellow water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable active comfort insole
Specifications
Color Green, Yellow
Material Cordura
Size 52
Toe Material Steel
Certifications/Compliance (Europe) EN ISO 20345: 2012 S3 SRC, ESD class 1 according EN 61340
Gender Men's
Ankle Style Low-cut
Waterproof Water Resistant
Product Type S1P SRC ESD