Learn More
ABEBA™ Anatom Green/Yellow Safety Sandals S1P
Product Code. 17161155
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 17161155
Brand: ABEBA™ 2283/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Robust, durable sandals made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
  • Comply to EN ISO 20345: 2012, S1P, SRC
  • Slip resistant PU double density outsole
  • Green/yellow water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable ACC wave insole
Specifications
Color Green, Yellow
Material Cordura
Size 36
Toe Material Steel
Certifications/Compliance (Europe) EN ISO 20345: 2012 S1P SRC
Gender Men's
Ankle Style Sandals
Waterproof Water Resistant
Product Type S1P SRC