Learn More
ABEBA™ Anatom Green/Yellow Safety Sandale S1P ESD
Product Code. 17191235
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 17191235
Brand: ABEBA™ 32283/44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Green/yellow sandals made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
  • Comfortable, robust, durable sandals
  • Comply to EN ISO 20345: 2012, S1P, SRC
  • ESD according EN 61340 class climatic 1
  • Slip resistant PU double density outsole
  • Green/yellow water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable active comfort insole
Specifications
Color Green, Yellow
Material Cordura
Size 44
Toe Material Steel
Certifications/Compliance (Europe) EN ISO 20345: 2012 S1P SRC, EN 61340 ESD class climatic 1
Gender Men's
Ankle Style Sandals
Waterproof No
Product Type S1P SRC ESD