Learn More
ABEBA™ Anatom Black Safety Lace Up Shoes S3
Product Code. 16328071
Click to view available options
Size:
49
50
51
52
Unit Size:
1 pair
Product Code. 16328071
Brand: ABEBA™ 32256/49
Only null left
Add to basket
Add to basket
ABEBA™ Anatom Black Safety Lace Up Shoes S3 are made of water resistant smooth leather with slip resistant PU2D outsole.
  • Comfort, robustness, durability
  • Comply to EN ISO 20345 : 2012, S3, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU double density outsole
  • Water resistant smooth black leather
  • Steel toe cap (200 joules)
  • Textile antipuncture midsole
  • Scratch-resistant coating in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable ACC Wave insole
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S3 SRC ESD
Closure Type Laces
Color Black
Eyelets Material Stainless steel
Liner Material Breathable silver ions lining
Material Leather
Size 49
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Polyurethane2D
Style Low-cut
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes