Learn More
ABEBA™ Anatom Black/Red Safety Shoes S3
Product Code. 17161125
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17161125
Brand: ABEBA™ 2238/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Robust, durable, black/red water and tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
  • Comfort, robustness, durability
  • Comply to EN ISO 20345: 2012, S3, SRC
  • Slip resistant PU double density outsole
  • Black/red water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable active comfort insole
Specifications
Certifications/Compliance EN ISO 20345: 2012, S3, SRC
Color Black, Red
Material Cordura
Size 36
Toe Material Steel
Gender Men's
Ankle Style Low-cut
Waterproof Yes
Product Type S3 SRC