Learn More
Abeba™ Anatom 32256 Shoes
Product Code. 15649547
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15649547 36 1 pair
15659547 37 1 pair
15669547 38 1 pair
15679547 39 1 pair
15689547 40 1 pair
15699547 41 1 pair
15609557 42 1 pair
15619557 43 1 pair
15629557 44 1 pair
15639557 45 1 pair
15649557 46 1 pair
15659557 47 1 pair
15669557 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15649547 Supplier Abeba™ Supplier No. 32256/36
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Scratch-resistant coating in toe and heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRC
Color Black
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32256
Toe Material Steel