Learn More
Abeba™ Anatom 32256 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marke: Abeba™ 32256/41
Additional Details : Weight : 1.40000kg
Beschreibung
- CE EN ISO 20345:2011 S3 SRC
- Smooth leather
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Scratch-resistant coating in toe and heel area
Spezifikation
CE EN ISO 20345:2011 S3 SRC | |
Leather | |
Slip-resistant | |
32256 |
Black | |
41 | |
Polyurethane | |
Steel |