Learn More
Abeba™ Anatom 32189 Shoes
Product Code. 15699497
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15699497
Brand: Abeba™ 32189/43
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Adjustable instep area
  • velcro fastening
  • Reflective material
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Black
Material Velour
Size 43
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32189
Toe Material Steel