missing translation for 'onlineSavingsMsg'
Learn More

Anastrozole Diamide, TRC

Product Code. 30339037
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339037 10 mg 10mg
30334718 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30339037 Supplier Toronto Research Chemicals Supplier No. TRCA63731510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anastrozole Diamide
CAS 120512-04-1
Molecular Formula C17 H23 N5 O2
Quantity 10 mg
Synonym 1,3-Benzenediacetamide, α1,α1,α3,α3-tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, 1,3-Benzenediacetamide, α,α,α',α'-tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)- (9CI), 2,2'-[5-(1H-1,2,4-Triazol-1-yl)methylbenzene-1,3-diyl]bis(2-methylpropanamide), 2-[3-(2-Amino-1,1-dimethyl-2-oxo-ethyl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanamide
InChI Formula InChI=1S/C17H23N5O2/c1-16(2,14(18)23)12-5-11(8-22-10-20-9-21-22)6-13(7-12)17(3,4)15(19)24/h5-7,9-10H,8H2,1-4H3,(H2,18,23)(H2,19,24)
SMILES CC(C)(C(=O)N)c1cc(Cn2cncn2)cc(c1)C(C)(C)C(=O)N
IUPAC Name 2-[3-(1-amino-2-methyl-1-oxopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanamide
Molecular Weight (g/mol) 329.4
Formula Weight 329.1852
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.