missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC2 Antibody (OTI1A6), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71860PECY55
This item is not returnable.
View return policy
Description
ANAPC2 Monoclonal specifically detects ANAPC2 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
| ANAPC2 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ANAPC2 | |
| Full length recombinant protein of human ANAPC2 (NP_037498) produced in HEk293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2a |
| Flow Cytometry | |
| OTI1A6 | |
| Flow Cytometry | |
| anaphase promoting complex subunit 2, anaphase-promoting complex subunit 2, APC2KIAA1406Cyclosome subunit 2, RP11-350O14.5 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 29882.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction