missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC2 Antibody (OTI1A6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71860DL594
This item is not returnable.
View return policy
Description
ANAPC2 Monoclonal antibody specifically detects ANAPC2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ANAPC2 | |
| Monoclonal | |
| DyLight 594 | |
| anaphase promoting complex subunit 2, anaphase-promoting complex subunit 2, APC2KIAA1406Cyclosome subunit 2, RP11-350O14.5 | |
| Full length recombinant protein of human ANAPC2 (NP_037498) produced in HEk293 cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 29882 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction