missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC13 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76261R
This item is not returnable.
View return policy
Description
ANAPC13 Polyclonal antibody specifically detects ANAPC13 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ANAPC13 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 550 | |
| anaphase promoting complex subunit 13, anaphase-promoting complex subunit 13, APC13SWM1, cyclosome subunit 13, Cylosome subunit 13, DKFZP566D193, homolog of yeast Swm1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human APC13. The immunogen is located within the last 50 amino acids of APC13. Amino Acid Squence: PLNELPEPEQDNGGTTE | |
| 0.1 mL | |
| Cell Biology | |
| 25847 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction