missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Anandamide - d4
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Anandamide - d4 is a deuterated anandamide . Used as an internal standard for the quantification of AEA by GC- or LC-mass spectrometry.Anandamide also available.
Specifications
Specifications
| Chemical Name or Material | N-(2-Hydroxyethyl-1,1,2,2-d4)-5Z,8Z,11Z,14Z-eicosatetraenamide |
| CAS | 946524-40-9 |
| Quantity | 1 mg |
| Target | Stable Isotopically Labeled Compounds |
| Molecular Formula | C22H33D4NO2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction