Learn More
Kern Sohn Analytical Balance ADJ
Product Code. 17351543
Encompass_Preferred
Click to view available options
Capacity:
120 g
210 g
Model:
ADJ
ADJ Compact
Unit Size:
Each
Product Code. 17351543
Brand: Kern Sohn ADJ 1004
Leading analytical balance with internal adjustment.
  • Automatic internal adjustment in the case of a change in temperature = 2 °C or time-controlled every 3 h, guarantees high degree of accuracy and makes the balance independent of its location of use
  • ADJ 600-C3: Compact, space-saving carat balance with a readout of 0.001 ct and a weighing range of 600 ct. The high level of accuracy saves hard cash wherever you are weighing valuable precious stones
  • Level indicator and levelling feet for precise levelling of the scale, fitted as standard, to give the most accurate weighing result
  • Large glass draught shield with 3 sliding doors for easy access to the items being weighed.
  • Compact size, practical for small spaces
  • Simple and convenient 6-key operation
Specifications
Capacity 120 g
Readability 0.1 mg
Linearity 0.4 mg
Model ADJ
Repeatability 0.2 mg
Electrical Requirements Universal Mains Adapter