Learn More
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Aspon | 3244-90-4 | 200μg/mL |
| Azinphos-ethyl | 2642-71-9 | 200μg/mL |
| Carbophenothion | 786-19-6 | 200μg/mL |
| Chlorfenvinfos | 470-90-6 | 200μg/mL |
| Chlorpyrifos-methyl | 5598-13-0 | 200μg/mL |
| Crotoxyphos | 7700-17-6 | 200μg/mL |
| Dichlorfenthion | 97-17-6 | 200μg/mL |
| Dicrotophos | 141-66-2 | 200μg/mL |
| Dimethoate | 60-51-5 | 200μg/mL |
| Dioxathion | 78-34-2 | 200μg/mL |
| Dyfonate (Fonofos) | 944-22-9 | 200μg/mL |
| Dylox | 52-68-6 | 200μg/mL |
| EPN | 2104-64-5 | 200μg/mL |
| Ethion | 563-12-2 | 200μg/mL |
| Ethyl parathion | 56-38-2 | 200μg/mL |
| Fenitrothion | 122-14-5 | 200μg/mL |
| Imidan | 732-11-6 | 200μg/mL |
| Leptophos | 21609-90-5 | 200μg/mL |
| Malathion | 121-75-5 | 200μg/mL |
| Monocrotophos | 6923-22-4 | 200μg/mL |
| Phosphamidon(33.4% E; 64.6% Z) | 13171-21-6 | 200μg/mL |
| TEPP | 107-49-3 | 200μg/mL |
| Terbufos | 13071-79-9 | 200μg/mL |
| Tetraethyldithio Pyrophosphate/Sulfotepp | 3689-24-5 | 200μg/mL |
| Zinophos | 297-97-2 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Analyte Mix |
| Name Note | SW-846 Method 8141 |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Method 8141 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | Hexane:Acetone |
| Quantity | 1 mL |
Safety and Handling
- ANALYTE MIX
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: n-hexane, acetone
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.