missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Analyte Mix C, High Level, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,1,1,2-Tetrachloroethane | 630-20-6 | 2000μg/mL |
| 1,1,1-Trichloroethane | 71-55-6 | 2000μg/mL |
| 1,1-Dichloropropene | 563-58-6 | 2000μg/mL |
| 1,2,3-Trichloropropane | 96-18-4 | 2000μg/mL |
| 1,2-Dibromoethane | 106-93-4 | 2000μg/mL |
| 1,2-Dichlorobenzene | 95-50-1 | 2000μg/mL |
| 1,3-Dichlorobenzene | 541-73-1 | 2000μg/mL |
| 1,3-Dichloropropane | 142-28-9 | 2000μg/mL |
| 2-Chlorotoluene | 95-49-8 | 2000μg/mL |
| 4-Chlorotoluene | 106-43-4 | 2000μg/mL |
| Bromobenzene | 108-86-1 | 2000μg/mL |
| Bromochloromethane | 74-97-5 | 2000μg/mL |
| Bromodichloromethane | 75-27-4 | 2000μg/mL |
| cis-1,2-Dichloroethene | 156-59-2 | 2000μg/mL |
| cis-1,3-Dichloropropene | 10061-01-5 | 2000μg/mL |
| Dibromomethane | 74-95-3 | 2000μg/mL |
| Ethylbenzene | 100-41-4 | 2000μg/mL |
| Isopropylbenzene | 98-82-8 | 2000μg/mL |
| n-Butylbenzene | 104-51-8 | 2000μg/mL |
| p-Xylene | 106-42-3 | 2000μg/mL |
| Styrene | 100-42-5 | 2000μg/mL |
| trans-1,3-Dichloropropene | 10061-02-6 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Analyte Mix C (High Level) |
| Name Note | USEPA Method 502.2, 524.2, 8260B |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Methods 502.2, 524.2, 8260, 8260B |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | Methanol-P and T |
| Quantity | 1 mL |
Safety and Handling
Product Identifier
- Analyte Mix C (High Level)
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : bromodichloromethane, 1,2,3-trichloropropane, isopropylbenzene, ethylbenzene, styrene, 1,2-dibromoet
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction