Learn More
Grant Instruments™ XUBA Analog Ultrasonic Bath
Provides a high standard of reliable and effective ultrasonic technology. Grant Instruments™ XUBA Analog Ultrasonic includes M2 cleaning solution, stainless steel basket and ABS plastic lid included as standard.
Brand: Grant Instruments™ XUBA1
Code : 57
Additional Details : Weight : 3.06000kg
Description
- Excellent entry level ultrasonic baths
- Temperature range ambient +5°C to 70°C on the XUBA3
- Fast, effective, efficient, easy and safe cleaning and processing of diverse instruments, components and solutions
- Supplied with stainless steel basket and ABS plastic lid as standard
- One bottle of M2 ultrasonic cleaning solution included as standard
- Control panel easy to operate even when wearing gloves
- Timer control from 0-15 minutes
Specifications
0 to 15 min. | |
230 V | |
1.5 L |
Safety and Handling
- Grant M2 ultrasonic solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : C9XYLATE, QUATERNARY FATTY AMINE ETHOXYLATE, BENZENESULFONIC ACID