Learn More
BINDER™ Analog Output 4-20 mA
Product Code. 17807128 Shop All BINDER Products
Click to view available options
For Use With (Application):
Temperature Monitoring
Unit Size:
Each
Product Code. 17807128
Brand: BINDER™ 80122295
Only null left
Add to basket
Add to basket
Analog output 4-20 mA - for temperature values (output not adjustable)
  • Accessory
Specifications
For Use With (Equipment) M E3.1 Drying and Heating Oven (M 56, M 115, M 260, M 720)
Material Aluminum
Type Analog Output
For Use With (Application) Temperature Monitoring
Amperage 4 to 20 mA