Learn More
TFA Dostmann Analog Kitchen Timer
Description
When it comes to cooking in minutes, this classic timer does a good job. With its functional design, it not only ensures the perfect breakfast egg, it also reminds you when the cake is in the oven or the pasta is in the pot. You can set up to 60 minutes with the mechanical winding mechanism. You are reliably reminded at the right time by an acoustic signal. Please be aware that the mechanical timer must be fully wound up before it will work.
- Windup mechanism, no battery required
- Continuously adjustable up to 60 min.
- Audio bell alarm after the time has elapsed
- Easy to read numbers
- Classic design
Specifications
Specifications
| Product Type | Mechanical Timer |
| Time Range | 0 to 60 min. |
| Alarms | Audible |
| Resolution | 1 min. |
| Display | Analog |
| Battery Type | None |
| Color | White |
| Weight (Metric) | 82 g |