missing translation for 'onlineSavingsMsg'
Learn More

Anacardic Acid, MedChemExpress

Product Code. 16473936
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473936 25 mg 25mg
16469248 10 mg 10mg
16441668 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473936 Supplier Medchem Express Supplier No. HYN2020
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Anacardic Acid, extracted from cashew nut shell liquid, is a histone acetyltransferase inhibitor, inhibits HAT activity of p300 and PCAF, with IC50s of ∼8.5 μM and ∼5 μM, respectively.

Anacardic Acid, extracted from cashew nut shell liquid, is a histone acetyltransferase inhibitor, inhibits HAT activity of p300 and PCAF, with IC50s of ∼8.5 μM and ∼5 μM, respectively.

Specifications

Chemical Name or Material Anacardic Acid
CAS 16611-84-0
Color White
Purity Grade Notes Research
Molecular Formula C22H36O3
Quantity 25 mg
Synonym Hydroginkgolic acidGinkgolic Acid C15:0
Solubility Information DMSO : 100 mg/mL (286.93 mM; Need ultrasonic)
SMILES O=C(O)C1=C(CCCCCCCCCCCCCCC)C=CC=C1O
Molecular Weight (g/mol) 348.52
Formula Weight 348.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.