missing translation for 'onlineSavingsMsg'
Learn More

Anacardic Acid, MedChemExpress

Product Code. 16441668
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16441668

Brand: Medchem Express HYN2020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anacardic Acid, extracted from cashew nut shell liquid, is a histone acetyltransferase inhibitor, inhibits HAT activity of p300 and PCAF, with IC50s of ∼8.5 μM and ∼5 μM, respectively.

Anacardic Acid, extracted from cashew nut shell liquid, is a histone acetyltransferase inhibitor, inhibits HAT activity of p300 and PCAF, with IC50s of ∼8.5 μM and ∼5 μM, respectively.

Specifications

Chemical Name or Material Anacardic Acid
CAS 16611-84-0
Color White
Purity Grade Notes Research
Molecular Formula C22H36O3
Quantity 5 mg
Synonym Hydroginkgolic acidGinkgolic Acid C15:0
Solubility Information DMSO : 100 mg/mL (286.93 mM; Need ultrasonic)
SMILES O=C(O)C1=C(CCCCCCCCCCCCCCC)C=CC=C1O
Molecular Weight (g/mol) 348.52
Formula Weight 348.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.