missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anabasine, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Description
Anabasine ((S)-Anabasine) is an alkaloid that found as a minor component in tobacco (Nicotiana). Anabasine is a botanical pesticide nicotine, acts as a full agonist of nicotinic acetylcholine receptors (nAChRs). Anabasine induces depolarization of TE671 cells endogenously expressing human fetal muscle-type nAChRs (EC50=0.7 μM).
Specifications
Specifications
| Chemical Name or Material | Anabasine |
| Physical Form | Liquid |
| CAS | 494-52-0 |
| Color | Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H14N2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | (S)-Anabasine (+)-Anabasine |
| Solubility Information | DMSO : 100 mg/mL (616.41 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction