missing translation for 'onlineSavingsMsg'
Learn More

ANA-12, MedChemExpress

Product Code. 16495208
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495208 10 mg 10mg
16451427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495208 Supplier Medchem Express Supplier No. HY12497
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ANA-12 is a potent and selective TrkB antagonist with IC50s of 45.6 nM and 41.1 μM for the high and low affinity sites, respectively.

ANA-12 is a potent and selective TrkB antagonist with IC50s of 45.6 nM and 41.1 μM for the high and low affinity sites, respectively.

Specifications

Chemical Name or Material ANA-12
CAS 219766-25-3
Color White
Purity Grade Notes Research
Molecular Formula C22H21N3O3S
Quantity 10 mg
Solubility Information DMSO : 9.09 mg/mL (22.31 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(C1=CC2=CC=CC=C2S1)NC3=CC=CC=C3C(NC4C(NCCCC4)=O)=O
Molecular Weight (g/mol) 407.49
Formula Weight 407.49
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.