missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550605
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total AMZ1 protein.
Zinc metalloprotease. Exhibits aminopeptidase activity against neurogranin in vitro. Does not hydrolyze angiotensin-2.
Specifications
| AMZ1 | |
| Polyclonal | |
| Unconjugated | |
| Amz1 | |
| 5330426I05; 6530401C20Rik; AMZ1; archaelysin family metallopeptidase 1; archaemetzincin-1; archeobacterial metalloproteinase-like 1; archeobacterial metalloproteinase-like protein 1; KIAA1950; metalloproteinase-like protein; RGD1311314 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 155185 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q400G9 | |
| Amz1 | |
| Synthetic peptide corresponding to residues near the C terminal of human archaelysin family metallopeptidase 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction