missing translation for 'onlineSavingsMsg'
Learn More

amyloid P-IN-1, MedChemExpress

Product Code. 16443847
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443847 50 mg 50mg
16481859 1 mg 1mg
16477898 10 mg 10mg
16417716 100 mg 100mg
16410368 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16443847 Supplier Medchem Express Supplier No. HY19771
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress amyloid P-IN-1 is used in the research of diseases or disorders wherein depletion of serum amyloid P component (SAP), including amyloidosis, Alzheimer's disease, type 2 diabetes mellitus and osteoarthritis.

amyloid P-IN-1 is used in the research of diseases or disorders wherein depletion of serum amyloid P component (SAP), including amyloidosis, Alzheimer's disease, type 2 diabetes mellitus and osteoarthritis.

Specifications

Chemical Name or Material amyloid P-IN-1
Physical Form Solid
CAS 1819986-22-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C30H44N2O14
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 34 mg/mL (51.78 mM)
SMILES O=C(OCOC(OC1CCOCC1)=O)[C@H]2CCCN2C(CCCCC(N3[C@@H](C(OCOC(OC4CCOCC4)=O)=O)CCC3)=O)=O
Molecular Weight (g/mol) 656.68
Formula Weight 656.68
Percent Purity 99.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.