Learn More
Thermo Scientific Chemicals Amyloid beta (1-16)
For cellular and molecular biology applications
Brand: Thermo Scientific™ J66562.#0
Code : NEW
Additional Details : CAS Number : 131580-10-4 Weight : 0.00005kg
Description
Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer’s disease and neurodegeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
Amyloid beta (1-16) | |
RUO - research use only |
-30°C to -10°C | |
1955.00 | |
5 mg | |
Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys |
RUO – Research Use Only