missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress β-Amyloid (31-35)

Product Code. 16482158
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16482158

Brand: MedChemExpress HYP1517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress β-Amyloid (31-35) is the shortest sequence of native Amyloid-β peptide that retains neurotoxic activity.

Specifications

Purity 99.72%
CAS 149385-65-9
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Neuroscience-Neurodegeneration
Molecular Weight (g/mol) 545.74 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : 5 mg/mL (9.16 mM∣ultrasonic and adjust pH to 7 with HCl)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.