missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress β-Amyloid (1-42), human TFA

Product Code. 16432889
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432889 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432889 Supplier MedChemExpress Supplier No. HYP1363
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress β-Amyloid (1-42), human TFA (Amyloid β-Peptide (1-42) (human) TFA) is a 42-amino acid peptide which plays a key role in the pathogenesis of Alzheimer disease.

Specifications

Purity 96.23%
CAS 107761-42-2
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
For Use With (Application) Neuroscience-Neurodegeneration
Molecular Weight (g/mol) 4628.06 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 33.33 mg/mL (7.20 mM∣Need ultrasonic)
Synonym Amyloid β-Peptide (1-42) (human) TFA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.