missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amylin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102889
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Amylin.
Islet, or insulinoma, amyloid polypeptide is commonly found in pancreatic islets of patients suffering diabetes mellitus type II, or harboring an insulinoma. While the association of amylin with the development of type II diabetes has been known for some time, a direct causative role for amylin has been harder to establish. Studies suggest that amylin, like the related beta-amyloid (Abeta) associated with Alzheimer's disease, can induce apoptotic cell-death in particular cultured cells, an effect that may be relevant to the development of type II diabetes.
Specifications
| Amylin | |
| Polyclonal | |
| Unconjugated | |
| IAPP | |
| Amylin; amylin peptide; DAP; Diabetes-associated peptide; IAP; Iapp; Insulinoma amyloid peptide; islet amyloid polypeptide; Islet amyloid polypeptide (diabetes-associated peptide; amylin); Rabbit Anti-Amylin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15874, 24476, 3375 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10997, P12968, P12969 | |
| IAPP | |
| A synthesized peptide derived from human IAPP(Accession P10997), corresponding to amino acid residues F9-I59. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction