missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amylin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69210
This item is not returnable.
View return policy
Description
Amylin Polyclonal specifically detects Amylin in Human samples. It is validated for Western Blot.
Specifications
| Amylin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin) | |
| Rabbit | |
| 4 kDa | |
| 100 μL | |
| Angiogenesis, Apoptosis, Cancer | |
| 3375 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P10997 | |
| IAPP | |
| Synthetic peptides corresponding to IAPP (islet amyloid polypeptide) The peptide sequence was selected from the N terminal of IAPP (NP_000406). Peptide sequence MGILKLQVFLIVLSVALNHLKATPIESHQVEKRKCNTATCATQRLANFLV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Bovine: 82%. | |
| Human, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction