missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amylase alpha Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15845-1-AP-20UL
This item is not returnable.
View return policy
Description
15845-1-AP can recognize both AMY1 and AMY2.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.Specifications
| Amylase alpha | |
| Polyclonal | |
| Unconjugated | |
| AMY2A | |
| AMY2A, Amylase, alpha, Pancreatic alpha amylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100043207, 279, 545562 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04746 | |
| AMY2A, Amy2a1, Amy2b | |
| Amylase, alpha Fusion Protein Ag8605 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction