missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMXT-1501 tetrahydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
AMXT-1501 tetrahydrochloride is an orally active polyamine transport inhibitor. AMXT1501 blocks tumor growth in immunocompetent mice but not in athymic nude mice lacking T cells. Combination of DFMO and AMXT‐1501 induces caspase‐3 mediated apoptosis in NB cell lines.
Specifications
Specifications
| Chemical Name or Material | AMXT-1501 tetrahydrochloride |
| Physical Form | Solid |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C32H72Cl4N6O2 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | H2O : 83.33 mg/mL (116.58 mM; Need ultrasonic) |
| SMILES | CCCCCCCCCCCCCCCC(NCCCC[C@@H](N)C(NCCCNCCCCNCCCN)=O)=O.Cl.Cl.Cl.Cl |
| Molecular Weight (g/mol) | 714.77 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction