missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMTN Antibody (OTI2D9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72507JF646
This item is not returnable.
View return policy
Description
AMTN Monoclonal specifically detects AMTN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AMTN | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AMTN | |
| Full length recombinant protein of human AMTN (NP_997722) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2D9 | |
| Western Blot, Immunohistochemistry | |
| amelotin, MGC148132, MGC148133, RSTI689, UNQ689 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 401138.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction