missing translation for 'onlineSavingsMsg'
Learn More

Amthamine dihydrobromide, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001111
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001111 10 mg 10mg
18054272 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001111 Supplier Tocris Bioscience Supplier No. 0668/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Highly selective standard H2 agonist

Highly selective H2 agonist, slightly more potent than histamine itself. Only a weak antagonist at H3 and has no activity at H1 receptors. Induces vasodilation of cerebral arteries and decreases myogenic tone in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142457-00-9
Molecular Formula C6H13Br2N3S
Molecular Weight (g/mol) 319.059
InChI Key XFXNNOPUDSFVJE-UHFFFAOYSA-N
Synonym amthamine dihydrobromide, 5-thiazoleethanamine,2-amino-4-methyl, amthamine dihydrobromide hplc , solid, 2-amino-4-methyl-5-thiazoleethanamine dihydrobromide, 2-amino-5-2-aminoethyl-4-methylthiazole dihydrobromide, 5-2-aminoethyl-4-methyl-1,3-thiazol-2-amine-hydrogen bromide 1/2
PubChem CID 16218912
IUPAC Name 5-(2-aminoethyl)-4-methyl-1,3-thiazol-2-amine;dihydrobromide
SMILES CC1=C(SC(=N1)N)CCN.Br.Br

Specifications

Quantity 10 mg
Formula Weight 319.06
Percent Purity >99%
Chemical Name or Material Amthamine dihydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.