missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMSH/STAMBP Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33217-20ul
This item is not returnable.
View return policy
Description
AMSH/STAMBP Monoclonal antibody specifically detects AMSH/STAMBP in Human,Monkey samples. It is validated for ELISA,Western Blot
Specifications
| AMSH/STAMBP | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| AMSHAssociated molecule with the SH3 domain of STAM, EC 3.1.2.15, EC 3.4.19.-, Endosome-associated ubiquitin isopeptidase, MGC126516, MGC126518, STAM binding protein, STAM-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human AMSH/STAMBP (O95630).,, Sequence:, FPKAEELKAELLKRYTKEYTEYNEEKKKEAEELARNMAIQQELEKEKQRVAQQKQQQLEQEQFHAFEEMIRNQELEKERLKIVQEFGKVDPGLGGPLVPDL | |
| 20 μL | |
| Cell Cycle and Replication | |
| 10617 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction