missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amrinone, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Amrinone |
| CAS | 60719-84-8 |
| Molecular Formula | C10 H9 N3 O |
| Quantity | 50 mg |
| Synonym | [3,4'-Bipyridin]-6(1H)-one, 5-amino-, 5-Amino[3,4'-bipyridin]-6(1H)-one, 3-Amino-5-(4-pyridinyl)-1,2-dihydro-2-pyridone, 5-Amino-1,6-dihydro-6-oxo-[3,4'-bipyridine], AWD 08-250, Amrinone, Cartonic, Cordemcura, Inamrinone, Vesistol, Win 40680, Wincoram |
| InChI Formula | InChI=1S/C10H9N3O/c11-9-5-8(6-13-10(9)14)7-1-3-12-4-2-7/h1-6H,11H2,(H,13,14) |
| SMILES | NC1=CC(=CNC1=O)c2ccncc2 |
| IUPAC Name | 3-amino-5-pyridin-4-yl-1H-pyridin-2-one |
| Molecular Weight (g/mol) | 187.2 |
| Formula Weight | 187.0746 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction