missing translation for 'onlineSavingsMsg'
Learn More

Ampyrone, MedChemExpress

Product Code. 16452339
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452339 Supplier Medchem Express Supplier No. HYB1398
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ampyrone is a reagent for glucose determination in the presence of peroxidase and phenol.

Ampyrone is a reagent for glucose determination in the presence of peroxidase and phenol.

Specifications

Chemical Name or Material Ampyrone
CAS 83-07-8
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C11H13N3O
Quantity 1 g
Synonym 4-Aminoantipyrine
Solubility Information DMSO : ≥ 50 mg/mL (246.01 mM) ∣H2O : 50 mg/mL (246.01 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=CC=C2)N(C)C(C)=C1N
Molecular Weight (g/mol) 203.24
Formula Weight 203.24
Percent Purity 98.72%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.