missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter AMPure XP Beads for DNA Cleanup

Product Code. 31723140 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Capacity:
5 mL
60 mL
450 mL
Unit Size:
450mL
5mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
31723140 5 mL 5mL
31723106 60 mL 60mL
31723123 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Refer to SDS
Product Code. 31723140 Supplier Beckman Coulter Supplier No. A63880
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Refer to SDS

The AMPure XP reagent, part of our SPRI reagent family, is a highly efficient, easily automated PCR purification system that delivers superior quality DNA with no salt carryover.

  • Requiring no centrifugation or filtration, AMPure XP reagents can be easily used in manual and automated 96- or 384-well formats.
TRUSTED_SUSTAINABILITY

Specifications

Capacity 5 mL
Isolation Technology Magnetic Bead
Product Type DNA Cleanup Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.