missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals AMPSO, 98+%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.
Solubility
Soluble in water (333.33 mg/ml, clear to slightly hazy colorless solution).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.
Solubility
Soluble in water (333.33 mg/ml, clear to slightly hazy colorless solution).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 215°C to 225°C |
| Color | White |
| Physical Form | Crystals |
| Quantity | 25 g |
| Beilstein | 5929916 |
| Solubility Information | Soluble in water (333.33mg/ml,clear to slightly hazy colorless solution). |
| Formula Weight | 227.28 |
| Percent Purity | ≥98% |
| Chemical Name or Material | AMPSO |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction